missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase-3 Antibody (31A1067), PerCP - (Pro and Active), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56708PCP
This item is not returnable.
View return policy
Description
Caspase-3 Monoclonal antibody specifically detects Caspase-3 in Human, Mouse, Rat, Porcine, Chicken, Chinese Hamster, Mammalia samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Hematoxylin and Eosin Stain, Electron Microscopy, Immunohistochemistry Free-Floating, CyTOF, Gene Knock-Out, KnockDown
Specifications
| Caspase-3 | |
| Monoclonal | |
| PerCP | |
| Apopain, apoptosis-related cysteine protease, CASP-3, caspase 3, apoptosis-related cysteine peptidase, caspase-3, CPP-32, CPP32B, CPP32SREBP cleavage activity 1, Cysteine protease CPP32, EC 3.4.22, EC 3.4.22.56, PARP cleavage protease, procaspase3, Protein Yama, SCA-1, Yama | |
| Full-length recombinant human caspase-3 protein was used as immunogen. The antibody recognizes an epitope in the large domain subunit of Caspase-3. As such it will recognize pro Caspase-3 and the large subunit cleavage fragment. | |
| 0.1 mL | |
| Alzheimers Research, Apoptosis, Cancer, Caspases, Cell Biology, Lipid and Metabolism, Neurodegeneration, Neuroscience, Signal Transduction | |
| 836 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| 31A1067 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Chicken | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction