Learn More
Caspase-3 Antibody (3H12) - Active, Pro, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26462-100ul
 View more versions of this product
Product Code. 30023747
  • € 533.00 / 100µL
Description
Caspase-3 Monoclonal antibody specifically detects Caspase-3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Caspase-3
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Western Blot, ELISA, Immunohistochemistry
3H12
Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200
Apopain, apoptosis-related cysteine protease, CASP-3, caspase 3, apoptosis-related cysteine peptidase, caspase-3, CPP-32, CPP32B, CPP32SREBP cleavage activity 1, Cysteine protease CPP32, EC 3.4.22, EC 3.4.22.56, PARP cleavage protease, procaspase3, Protein Yama, SCA-1, Yama
A synthesized peptide derived from Human Caspase-3 (Pro and Active) [UniProt P42574]
100 μL
Alzheimers Research, Apoptosis, Cancer, Caspases, Cell Biology, Lipid and Metabolism, Neurodegeneration, Neuroscience, Signal Transduction
836
Store at -20 to -70C. Avoid freeze-thaw cycles.