Learn More
Caspase 3/p17/p19 Rabbit anti-Human, CoraLite 594, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL594-19677-100UL
Description
The Caspase 3/p17/p19 antibody from Proteintech is a rabbit polyclonal antibody to a peptide of human Caspase 3/p17/p19. This antibody recognizes human, mouse, rat antigen. The Caspase 3/p17/p19 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™594.
Specifications
| Caspase 3/p17/p19 | |
| Polyclonal | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat |
| Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| CoraLite 594 | |
| 100 μL | |
| 836 | |
| IgG |