missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase 5 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98202MFV450
This item is not returnable.
View return policy
Description
Caspase 5 Polyclonal antibody specifically detects Caspase 5 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Caspase 5 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| CASP-5, caspase 5, apoptosis-related cysteine peptidase, caspase 5, apoptosis-related cysteine protease, caspase-5, EC 3.4.22, EC 3.4.22.58, ICE(rel)III, ICE(rel)-III, ICEREL-III, ICH3, ICH-3, MGC141966, Protease ICH-3, Protease TY, TY protease | |
| Produced in rabbits immunized with E. coli-derived Human Caspase 5 fragment. | |
| 0.1 mL | |
| Apoptosis, Cancer, Caspases | |
| 838 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction