missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Caspase 5 Antibody (SR3060), Novus Biologicals™

Rabbit Monoclonal Antibody

€ 316.00 - € 382.00

Specifications

Antigen Caspase 5
Clone SR3060
Dilution Western Blot 1:1000-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50
Applications Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30041634
View Documents
Novus Biologicals™
NBP3-27741-50ul
50 μL
€ 316.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30041512
View Documents
Novus Biologicals™
NBP3-27741-100ul
100 μL
€ 382.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Caspase 5 Monoclonal antibody specifically detects Caspase 5 in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

Caspase 5
Western Blot 1:1000-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50
Monoclonal
Purified
RUO
Human
CASP-5, caspase 5, apoptosis-related cysteine peptidase, caspase 5, apoptosis-related cysteine protease, caspase-5, EC 3.4.22, EC 3.4.22.58, ICE(rel)III, ICE(rel)-III, ICEREL-III, ICH3, ICH-3, MGC141966, Protease ICH-3, Protease TY, TY protease
A synthesized peptide derived from human Caspase 5 (Uniprot #: P51878)
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
SR3060
Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence
Unconjugated
Rabbit
Apoptosis, Cancer, Caspases
PBS (pH 7.4), 150mM NaCl, 50% glycerol
838
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.