missing translation for 'onlineSavingsMsg'
Learn More

Caspase-6 Antibody [DyLight 755], Novus Biologicals Biologicals™

Product Code. 30499262 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30499262 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30499262 Supplier Novus Biologicals Supplier No. NBP327875IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Caspase-6 Polyclonal antibody specifically detects Caspase-6 in Human samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Caspase-6
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias Apoptotic protease Mch-2, CASP-6, caspase 6, apoptosis-related cysteine peptidase, caspase 6, apoptosis-related cysteine protease, caspase-6, EC 3.4.22, MCH2EC 3.4.22.59
Host Species Rabbit
Immunogen Caspase-6 antibody was raised against a 15 amino acid synthetic peptide from near the center of human Caspase-6. The immunogen is located within amino acids 160 - 210 of Caspase-6.
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Caspases, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 839
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.