missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Caspase-6 Monoclonal antibody specifically detects Caspase-6 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | Caspase-6 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:2000, ELISA Recommended starting concentration is 1 μg/mL |
| Formulation | PBS (pH 7.3), 50% glycerol, 0.05% BSA |
| Gene Alias | Apoptotic protease Mch-2, CASP-6, caspase 6, apoptosis-related cysteine peptidase, caspase 6, apoptosis-related cysteine protease, caspase-6, EC 3.4.22, MCH2EC 3.4.22.59 |
| Host Species | Rabbit |
| Immunogen | A synthesized peptide derived from human Caspase-6. |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?