Learn More
Invitrogen™ Caspase 6 Monoclonal Antibody (M378)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA547700
Product Code. 17846978
  • € 484.00 / 100µL
Description
The antibody detects a 34 kDa* protein corresponding to the molecular weight of Caspase-6 on SDS-PAGE immunoblots of human Jurkat, K562, and A431 cells.
Caspases are cysteine proteases, expressed as inactive precursors, that mediate apoptosis by proteolysis of specific substrates. Caspases have the ability to cleave after aspartic acid residues. There are two classes of caspases involved in apoptosis; initiators (activation by receptor cluster) and effectors (activation by mitochondrial permeability transition). Proapoptotic signals autocatalytically activate initiator caspases, such as Caspase-8 and Caspase 9. Activated initiator caspases then process effector caspases, such as Caspase-3 and Caspase-7, which in turn cause cell collapse.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Caspase 6
Monoclonal
0.25 mg/mL
PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
P55212
Casp6
Clone (M378) was generated from a peptide containing amino acid residues in the C-terminal region of human Caspase-6.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
M378
Unconjugated
Casp6
apoptotic protease MCH-2; CASP6; CASP-6; caspase 6; caspase 6, apoptosis-related cysteine peptidase; caspase 6, apoptosis-related cysteine protease; caspase-6; Caspase-6 subunit p11; Caspase-6 subunit p18; mCASP-6; MCH2
Mouse
Protein A
RUO
839
-20°C, Avoid Freeze/Thaw Cycles