missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Caspase-7 Antibody (MCH3-5), Novus Biologicals™

Product Code. 30067106 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30067106 25 μg 25µg
30066831 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30067106 Supplier Novus Biologicals™ Supplier No. NBP33081425ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Caspase-7 Monoclonal antibody specifically detects Caspase-7 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Caspase-7
Applications Western Blot
Classification Monoclonal
Clone MCH3-5
Conjugate Unconjugated
Formulation Phosphate buffered saline
Gene Alias apoptosis-related cysteine protease, Apoptotic protease Mch-3, CASP-7, caspase 7, apoptosis-related cysteine peptidase, caspase-7, EC 3.4.22, EC 3.4.22.60, ICE-like apoptotic protease 3, Lice2 alpha/beta/gamma
Host Species Mouse
Immunogen Hybridoma produced by the fusion of splenocytes from mice immunized with recombinant human Caspase-7 protein and mouse myeloma cells.
Purification Method Ammonium sulfate precipitation
Quantity 25 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Caspases, Cell Biology, Cellular Markers, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 840
Target Species Human
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.