missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase-8 Antibody (90A992), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56527AF488
This item is not returnable.
View return policy
Description
Caspase-8 Monoclonal specifically detects Caspase-8 in Human, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Caspase-8 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| AIS, androgen receptor, DHTRTFM, Dihydrotestosterone receptorHYSP1, HUMARA, NR3C4KD, Nuclear receptor subfamily 3 group C member 4, SMAX1SBMA, spinal and bulbar muscular atrophy | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This can be used for detection of the pro-form of Caspase-8. | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) | |
| 90A992 | |
| Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin | |
| Q14790 | |
| CASP8 | |
| The immunogen for this product falls between amino acids 360-385 of the human Caspase 8 protein. | |
| 0.1 mL | |
| Apoptosis, Cancer, Caspases, Cell Biology, Phospho Specific, Ubiquitin Proteasome Pathway | |
| 841 | |
| Human, Primate | |
| Purified |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering