missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase-8 Antibody (90A992), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56527AF647
This item is not returnable.
View return policy
Description
Caspase-8 Monoclonal specifically detects Caspase-8 in Human, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Caspase-8 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| AIS, androgen receptor, DHTRTFM, Dihydrotestosterone receptorHYSP1, HUMARA, NR3C4KD, Nuclear receptor subfamily 3 group C member 4, SMAX1SBMA, spinal and bulbar muscular atrophy | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This can be used for detection of the pro-form of Caspase-8. | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) | |
| 90A992 | |
| Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin | |
| Q14790 | |
| CASP8 | |
| The immunogen for this product falls between amino acids 360-385 of the human Caspase 8 protein. | |
| 0.1 mL | |
| Apoptosis, Cancer, Caspases, Cell Biology, Phospho Specific, Ubiquitin Proteasome Pathway | |
| 841 | |
| Human, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction