Learn More
TTC19 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-98556MFV450
Product Code. 18538353
  • € 428.00 / 0.10mL
Description
TTC19 Polyclonal antibody specifically detects TTC19 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
mFluor Violet 450 SE
Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation