missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Caspase-9 Monoclonal antibody specifically detects Caspase-9 in Human, Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Caspase-9 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | S09-4C4 |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA |
| Gene Alias | APAF-3APAF3, apoptotic protease activating factor 3, Apoptotic protease Mch-6, Apoptotic protease-activating factor 3, CASP-9, caspase 9, apoptosis-related cysteine peptidase, caspase-9, EC 3.4.22.62, ICE-LAP6CASPASE-9c, ICE-like apoptotic protease 6, MCH6apoptosis-related cysteine protease |
| Host Species | Rabbit |
| Immunogen | Recombinant protein of human Caspase-9 (Uniprot # P55211) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?