Learn More
Caspase 9/p35/p10 Rabbit anti-Human, CoraLite 555, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL555-10380-100UL
Product Code. 17571616
  • € 450.00 / 100µL
Description
The Caspase 9/p35/p10 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human Caspase 9/p35/p10. This antibody recognizes human, mouse, rat antigen. The Caspase 9/p35/p10 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™555.
Specifications
Specifications
Caspase 9/p35/p10
Polyclonal
Rabbit
Primary
Human, Mouse, Rat
Flow Cytometry (Intracellular Staining)
CoraLite 555
100 μL
842