missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Caspr2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528969
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji, mouse brain. Predicted reactivity: Mouse (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the neurexin family which functions in the vertebrate nervous system as cell adhesion molecules and receptors. This protein, like other neurexin proteins, contains epidermal growth factor repeats and laminin G domains. In addition, it includes an F5/8 type C domain, discoidin/neuropilin- and fibrinogen-like domains, thrombospondin N-terminal-like domains and a putative PDZ binding site. This protein is localized at the juxtaparanodes of myelinated axons, and mediates interactions between neurons and glia during nervous system development and is also involved in localization of potassium channels within differentiating axons. This gene encompasses almost 1.5% of chromosome 7 and is one of the largest genes in the human genome. It is directly bound and regulated by forkhead box protein P2 (FOXP2), a transcription factor related to speech and language development. This gene has been implicated in multiple neurodevelopmental disorders, including Gilles de la Tourette syndrome, schizophrenia, epilepsy, autism, ADHD and mental retardation.
Specifications
| Caspr2 | |
| Polyclonal | |
| Unconjugated | |
| CNTNAP2 | |
| 5430425M22Rik; AUTS15; CASPR2; CDFE; Cell recognition molecule Caspr2; Cntnap2; contactin associated protein-like 2; contactin-associated protein-like 2; DKFZp781D1846; homolog of Drosophila neurexin IV; KIAA0868; mKIAA0868; NRXN4; PTHSL1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26047, 500105, 66797 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q9CPW0, Q9UHC6 | |
| CNTNAP2 | |
| Recombinant protein encompassing a sequence within the Extracellular domain of human Caspr2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction