missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CASZ1 Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06281APC
This item is not returnable.
View return policy
Description
CASZ1 Polyclonal antibody specifically detects CASZ1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CASZ1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| castor, castor homolog 1, zinc finger, castor homolog 1, zinc finger (Drosophila), castor zinc finger 1, Castor-related protein, CST, dJ734G22.1, FLJ20321, Putative survival-related protein, SRG, SRGcst, survival related, survival-related, Zinc finger protein 693, zinc finger protein castor homolog 1, ZNF693, ZNF693FLJ12223 | |
| Produced in rabbits immunized with E. coli-derived Human CASZ1 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| APC | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 54897 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction