missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CAT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576916
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Carnitine acetyltransferase (CRAT) is a key enzyme in the metabolic pathway in mitochondria, peroxisomes and endoplasmic reticulum. CRAT catalyzes the reversible transfer of acyl groups from an acyl-CoA thioester to carnitine and regulates the ratio of acylCoA/CoA in the subcellular compartments. Different subcellular localizations of the CRAT mRNAs are thought to result from alternative splicing of the CRAT gene suggested by the divergent sequences in the 5' region of peroxisomal and mitochondrial CRAT cDNAs and the location of an intron where the sequences diverge. The alternatively splicing of this gene results in three distinct isoforms, one of which contains an N-terminial mitochondrial transit peptide, and has been shown to be located in mitochondria.
Specifications
| CAT1 | |
| Polyclonal | |
| Unconjugated | |
| CRAT | |
| amino acid transporter, cationic 1; ATRC1; AW107812; CARAT; Carnitine acetylase; carnitine acetyltransferase; Carnitine O-acetyltransferase; CAT; CAT1; CAT-1; Crat; Ecotropic retroviral leukemia receptor homolog; ecotropic retroviral receptor; ecotropic retrovirus receptor homolog; ERR; HCAT1; High affinity cationic amino acid transporter 1; REC1L; SLC7A1; solute carrier family 7 (cationic amino acid transporter, y+ system), member 1; solute carrier family 7 member 1; System Y+ basic amino acid transporter | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12908, 1384, 311849 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P43155, P47934, Q704S8 | |
| CRAT | |
| Recombinant full length Human CAT1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction