missing translation for 'onlineSavingsMsg'
Learn More

Catalase Antibody (OTI1B8), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-70342MFV500

Product Code. 18470604

  • € 830.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Catalase Monoclonal antibody specifically detects Catalase in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Catalase
Monoclonal
mFluor Violet 500 SE
50mM Sodium Borate
Mouse
Immunogen affinity purified
RUO
Primary
Human, Mouse, Rat, Primate, Monkey
Purified
Flow Cytometry
OTI1B8
Flow Cytometry
catalase, EC 1.11.1.6, MGC138422, MGC138424
Full length human recombinant protein of human CAT (NP_111743) produced in HEK293T cell.
0.1 mL
Cancer, Cellular Markers, Hypoxia, Lipid and Metabolism, Neuroscience, Peroxisome Markers, Signal Transduction
847
Store at 4°C in the dark.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.