missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Catenin alpha 1 Antibody (1G5), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54533F
This item is not returnable.
View return policy
Description
Catenin alpha 1 Monoclonal antibody specifically detects Catenin alpha 1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, CyTOF
Specifications
| Catenin alpha 1 | |
| Monoclonal | |
| FITC | |
| Alpha E-catenin, alpha-catenin, alpha-E-catenin, alphaE-catenin, Cadherin-associated protein, CAP102, catenin (cadherin-associated protein), alpha 1 (102kD), catenin (cadherin-associated protein), alpha 1, 102kDa, catenin alpha-1, FLJ36832, FLJ52416, Renal carcinoma antigen NY-REN-13 | |
| Human alpha catenin fusion protein (Uniprot: P35221) | |
| 100 μg | |
| Cancer, Cell Biology, Cellular Markers, Core ESC Like Genes, Stem Cell Markers | |
| 1495 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunofluorescence, CyTOF | |
| 1G5 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction