missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Catenin alpha 1 Antibody (1G5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43548
This item is not returnable.
View return policy
Description
Catenin alpha 1 Monoclonal antibody specifically detects Catenin alpha 1 in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| Catenin alpha 1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation | |
| Alpha E-catenin, alpha-catenin, alpha-E-catenin, alphaE-catenin, Cadherin-associated protein, CAP102, catenin (cadherin-associated protein), alpha 1 (102kD), catenin (cadherin-associated protein), alpha 1, 102kDa, catenin alpha-1, FLJ36832, FLJ52416, Renal carcinoma antigen NY-REN-13 | |
| This antibody was raised by immunizing Balb/c mice with a 90 kDa MBP/Catenin alpha 1 fusion protein purified on an amylose column. (Uniprot# P35221) | |
| 0.2 mg | |
| Cancer, Cell Biology, Cellular Markers, Core ESC Like Genes, Phospho Specific, Stem Cell Markers | |
| 1495 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 1G5 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction