missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin 6 Antibody (RM0157-3L27) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
€ 195.00 - € 510.00
Specifications
| Antigen | Cathepsin 6 |
|---|---|
| Clone | RM0157-3L27 |
| Concentration | LYOPH |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18087476
|
Novus Biologicals
NBP2-12423-0.025MG |
0.025 mg |
€ 195.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18058066
|
Novus Biologicals
NBP2-12423 |
0.1 mg |
€ 510.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cathepsin 6 Monoclonal specifically detects Cathepsin 6 in Mouse samples. It is validated for Western Blot.Specifications
| Cathepsin 6 | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| NP_067420 | |
| 58518 | |
| IgG2 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| RM0157-3L27 | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| 1600022N02Rik | |
| Cts6 | |
| Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title