missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin 6 Antibody (RM0157-3L27), DyLight 755, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12423IR
This item is not returnable.
View return policy
Description
Cathepsin 6 Monoclonal specifically detects Cathepsin 6 in Mouse samples. It is validated for Western Blot.
Specifications
| Cathepsin 6 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Cts6 | |
| Mouse recombinant Cathepsin 6 | |
| 0.1 mL | |
| 58518 | |
| Store at 4C in the dark. | |
| IgG2 |
| Western Blot | |
| RM0157-3L27 | |
| Western Blot | |
| 1600022N02Rik | |
| Rat | |
| Protein A or G purified | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction