missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin 7 Antibody (RM0158-4A12), Alexa Fluor™ 750, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12424AF750
This item is not returnable.
View return policy
Description
Cathepsin 7 Monoclonal specifically detects Cathepsin 7 in Mouse samples. It is validated for Western Blot.
Specifications
| Cathepsin 7 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Cts7 | |
| Mouse recombinant Cathepsin 7 | |
| 0.1 mL | |
| 56092 | |
| Store at 4°C in the dark. |
| Western Blot | |
| RM0158-4A12 | |
| Western Blot | |
| CTS1, Epcs24, Epcs71 | |
| Rat | |
| Protein A or G purified | |
| Primary | |
| Mouse | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction