missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin B Antibody (010), Alexa Fluor™ 594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90402AF594
This item is not returnable.
View return policy
Description
Cathepsin B Monoclonal specifically detects Cathepsin B in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Cathepsin B | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 10 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| APP secretase, APPS, cathepsin B, Cathepsin B1, CPSBamyloid precursor protein secretase, cysteine protease, EC 3.4.22, EC 3.4.22.1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Cathepsin B (P10605; Met1-Phe339). | |
| 0.1 mL | |
| Alzheimers Research, Cancer, Cellular Markers, Neurodegeneration, Neuroscience | |
| 1508 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction