missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin E Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98304H
This item is not returnable.
View return policy
Description
Cathepsin E Polyclonal specifically detects Cathepsin E in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Cathepsin E | |
| Polyclonal | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CATE, cathepsin E, EC 3.4.23, EC 3.4.23.34, erythrocyte membrane aspartic proteinase, slow-moving proteinase | |
| Produced in rabbits immunized with purified, recombinant Cathepsin E (NP_031825.2; Met1-Pro397). | |
| 0.1 mL | |
| Adaptive Immunity, Immunology, Innate Immunity, Lipid and Metabolism | |
| 1510 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HRP | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction