Learn More
Cathepsin E Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-98304MFV610
Product Code. 18536370
  • € 451.00 / 0.10mL
Description
Cathepsin E Polyclonal antibody specifically detects Cathepsin E in Mouse samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Immunohistochemistry (Paraffin)
ELISA, Immunohistochemistry-Paraffin
RUO
Primary
IgG
mFluor Violet 610 SE
0.1 mL
Adaptive Immunity, Immunology, Innate Immunity, Lipid and Metabolism