Learn More
Invitrogen™ Cathepsin E Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA547116
Description
This antibody detects human pro and active Cathepsin E in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 40% cross-reactivity with recombinant mouse Cathepsin E is observed and less than 2% cross-reactivity with recombinant human (rh) Cathepsin D and rhBACE is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Cathepsin E may have a role in immune function. It is most likely involved in the processing of antigenic peptides during MHC class II-mediated antigen presentation. It may also play a role in activation-induced lymphocyte depletion in the thymus, and in neuronal degeneration and glial cell activation in the brain.
Specifications
| Cathepsin E | |
| Polyclonal | |
| Unconjugated | |
| CTSE | |
| A430072O03Rik; C920004C08Rik; CATE; Cathepsin E; Cathepsin E form I; Cathepsin E form II; CE; CEA; CEB; CTSE; Ctsea; EC 3.4.23.34; erythrocyte membrane aspartic proteinase; slow-moving proteinase | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 1510 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P14091 | |
| CTSE | |
| Mouse myeloma cell line NS0-derived recombinant human Cathepsin E Ile54-Pro396. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.