missing translation for 'onlineSavingsMsg'
Learn More

Cathepsin F Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 3 publications

Brand:  Novus Biologicals NB100-1784

Product Code. 18086775

  • € 506.00 / 0.10mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Cathepsin F Polyclonal specifically detects Cathepsin F in Human samples. It is validated for Western Blot, Peptide ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cathepsin F
Polyclonal
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
cathepsin F, CATSFEC 3.4.22.41, EC 3.4.22
Goat
Affinity Purified
RUO
Primary
Cross reacts with Human. Expected from sequence similarity: Mouse, Rat and Dog.
Store at -20C. Avoid freeze-thaw cycles.
Western Blot, ELISA
0.5 mg/mL
Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:32000
NP_003784.2
CTSF
Peptide with sequence CGVNTMASSAVVD corresponding to C-Terminus according to NP_003784.2.
0.1 mg
Immunology
8722
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only