missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cathepsin F Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100423
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CathepsinF.
Cathepsins are papain family cysteine proteinases that represent a major component of the lysosomal proteolytic system. Cathepsins generally contain a signal sequence, followed by a propeptide and then a catalytically active mature region. The very long (251 amino acid residues) proregion of the cathepsin F precursor contains a C-terminal domain similar to the pro-segment of cathepsin L-like enzymes, a 50-residue flexible linker peptide, and an N-terminal domain predicted to adopt a cystatin-like fold. The cathepsin F proregion is unique within the papain family cysteine proteases in that it contains this additional N-terminal segment predicted to share structural similarities with cysteine protease inhibitors of the cystatin superfamily. This cystatin-like domain contains some of the elements known to be important for inhibitory activity. CTSF encodes a predicted protein of 484 amino acids which contains a 19 residue signal peptide. Cathepsin F contains five potential N-glycosylation sites, and it may be targeted to the endosomal/lysosomal compartment via the mannose 6-phosphate receptor pathway. The cathepsin F gene is ubiquitously expressed, and it maps to chromosome 11q13, close to the gene encoding cathepsin W.
Specifications
| Cathepsin F | |
| Polyclonal | |
| Unconjugated | |
| Ctsf | |
| AI481912; Cathepsin F; CATSF; CLN13; CTSF; HGNC:2531 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 361704, 56464, 8722 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9R013, Q9UBX1 | |
| Ctsf | |
| A synthesized peptide derived from human CTSF(Accession Q9UBX1), corresponding to amino acid residues D330-S380. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction