missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin G Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 224.00 - € 530.00
Specifications
| Antigen | Cathepsin G |
|---|---|
| Dilution | ELISA Recommended starting concentration is 1 μg/mL, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
| Applications | ELISA, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30228977
|
Novus Biologicals
NBP3-33319-20ul |
20 μL |
€ 224.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30230411
|
Novus Biologicals
NBP3-33319-100ul |
100 μL |
€ 530.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cathepsin G Monoclonal antibody specifically detects Cathepsin G in Human samples. It is validated for ELISA,Immunocytochemistry/ImmunofluorescenceSpecifications
| Cathepsin G | |
| ELISA, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| 1511 | |
| IgG | |
| Affinity purified |
| ELISA Recommended starting concentration is 1 μg/mL, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| cathepsin G, CathepsinG, CGCATG, EC 3.4.21, EC 3.4.21.20, MGC23078 | |
| A synthetic peptide corresponding to a sequence within amino acids 156-255 of human Cathepsin G (NP_001902.1).,, Sequence:, TDTLREVQLRVQRDRQCLRIFGSYDPRRQICVGDRRERKAAFKGDSGGPLLCNNVAHGIVSYGKSSGVPPEVFTRVSSFLPWIRTTMRSFKLLDQMETPL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title