missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin K Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-99073CL1
This item is not returnable.
View return policy
Description
Cathepsin K Polyclonal antibody specifically detects Cathepsin K in Human samples. It is validated for Western Blot
Specifications
| Cathepsin K | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| CoraFluor 1 | |
| cathepsin K, cathepsin K (pycnodysostosis), Cathepsin O, cathepsin O1, Cathepsin O2, Cathepsin X, CTS02, CTSO, CTSO1, CTSO2, EC 3.4.22, EC 3.4.22.38, MGC23107, PKND, PYCD | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human Cathepsin K | |
| 0.1 mL | |
| Cancer | |
| 1513 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction