missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin K, Mouse anti-Human, Clone: CTSK/2793, Novus Biologicals™
Mouse Monoclonal Antibody
€ 256.00 - € 489.00
Specifications
| Antigen | Cathepsin K |
|---|---|
| Clone | CTSK/2793 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18249877
|
Novus Biologicals
NBP2-79765-20UG |
20 μg |
€ 256.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18228887
|
Novus Biologicals
NBP2-79765-100UG |
100 μg |
€ 489.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cathepsin K Monoclonal specifically detects Cathepsin K in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
| Cathepsin K | |
| 0.2 mg/mL | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| 10 mM PBS with 0.05% BSA | |
| cathepsin K, cathepsin K (pycnodysostosis), Cathepsin O, cathepsin O1, Cathepsin O2, Cathepsin X, CTS02, CTSO, CTSO1, CTSO2, EC 3.4.22, EC 3.4.22.38, MGC23107, PKND, PYCD | |
| CTSK | |
| IgG1 κ | |
| Protein A or G purified |
| CTSK/2793 | |
| Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P43235 | |
| 1513 | |
| Recombinant fragment of human Cathepsin K protein (around aa 163-274) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title