Learn More
Cathepsin L Antibody (221), Allophycocyanin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-89528APC
Product Code. 18692841
  • € 607.00 / 0.10mL
Description
Cathepsin L Monoclonal antibody specifically detects Cathepsin L in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cathepsin L
Monoclonal
APC
PBS
Rabbit
Protein A purified
RUO
Primary
Human
Purified
Western Blot
221
Western Blot
cathepsin L, cathepsin L1, CATL, CTSLEC 3.4.22.15, EC 3.4.22, FLJ31037, Major excreted protein, MEP
This antibody was obtained from a rabbit immunized with purified, recombinant Human Cathepsin L (Accession#: NP_001903.1; Met1-Val333).
0.1 mL
Cancer, Cellular Markers
1514
Store at 4C in the dark.