missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin L Antibody (221), Janelia Fluor™ 646, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89528JF646
This item is not returnable.
View return policy
Description
Cathepsin L Monoclonal antibody specifically detects Cathepsin L in Human samples. It is validated for Western Blot
Specifications
| Cathepsin L | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 221 | |
| Western Blot | |
| cathepsin L, cathepsin L1, CATL, CTSLEC 3.4.22.15, EC 3.4.22, FLJ31037, Major excreted protein, MEP | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Cathepsin L (Accession#: NP_001903.1; Met1-Val333). | |
| 0.1 mL | |
| Cancer, Cellular Markers | |
| 1514 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction