missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin V Antibody [Janelia Fluor™ 635], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-26082JF635
This item is not returnable.
View return policy
Description
Cathepsin V Polyclonal antibody specifically detects Cathepsin V in Human samples. It is validated for ELISA
Specifications
| Cathepsin V | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 1515 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| Janelia Fluor 635 | |
| cathepsin L2, Cathepsin U, Cathepsin V, CATL2, CTSUcathepsin L2, preproprotein, CTSVMGC125957, EC 3.4.22, EC 3.4.22.43 | |
| Produced in rabbits immunized with purified, recombinant Human Cathepsin V (Accession#: NP_001324.2; Met1-Val334) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction