missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Cathepsin Z Antibody (05) [DyLight 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26880DL594
This item is not returnable.
View return policy
Description
Cathepsin Z Monoclonal antibody specifically detects Cathepsin Z in Human samples. It is validated for ELISA
Specifications
| Cathepsin Z | |
| Monoclonal | |
| DyLight 594 | |
| Cathepsin P, Cathepsin X, cathepsin Z, CTSX, EC 3.4.18.1, FLJ17088, preprocathepsin P | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Cathepsin Z (Uniprot#: Q9UBR2; Met1-Val303). | |
| 0.1 mL | |
| Cancer | |
| 1522 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 05 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction