missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin Z Antibody (S09-9J9), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 444.00 - € 680.00
Specifications
| Antigen | Cathepsin Z |
|---|---|
| Clone | S09-9J9 |
| Concentration | 0.3 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18610575
|
Novus Biologicals
NBP3-19594-100ul |
100 μg |
€ 680.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18603705
|
Novus Biologicals
NBP3-19594-50ul |
50 μg |
€ 444.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cathepsin Z Monoclonal antibody specifically detects Cathepsin Z in Human, Mouse samples. It is validated for Western BlotSpecifications
| Cathepsin Z | |
| 0.3 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| Cathepsin P, Cathepsin X, cathepsin Z, CTSX, EC 3.4.18.1, FLJ17088, preprocathepsin P | |
| Recombinant protein of human Cathepsin Z (Uniprot # Q9UBR2) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| S09-9J9 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 1522 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title