missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CATIP Antibody (OTI4B4), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72506AF750
This item is not returnable.
View return policy
Description
CATIP Monoclonal specifically detects CATIP in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| CATIP | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| CATIP | |
| Full length recombinant protein of human C2orf62 (NP_940961) produced in HEK293T cells. | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG2a |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI4B4 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| C2orf62, chromosome 2 open reading frame 62, hypothetical protein LOC375307, MGC50811 | |
| Mouse | |
| 0.1 mL | |
| 375307 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction