missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caveolin-1 Antibody (7C8), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-615AF647
This item is not returnable.
View return policy
Description
Caveolin-1 Monoclonal specifically detects Caveolin-1 in Human, Mouse, Rat, Sheep samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Caveolin-1 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| CAV, caveolae protein, 22kD, caveolin 1, caveolae protein, 22kDa, cell growth-inhibiting protein 32, MSTP085 | |
| Mouse | |
| 23 kDa | |
| 0.1 mL | |
| Cancer, Caveolae Markers, Cellular Markers, Cholesterol Metabolism, Hypoxia, Lipid and Metabolism, Lipid Droplets, Phospho Specific, Signal Transduction | |
| 857 | |
| Human, Mouse, Rat, Sheep | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 7C8 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| CAV1 | |
| Intracellular membrane protein-containing vesicles (containing GLUT4) from rat adipocytes | |
| Protein A purified | |
| RUO | |
| Primary | |
| This is specific for caveolin alpha and beta proteins. | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction