missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caveolin-2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 224.00 - € 326.00
Specifications
| Antigen | Caveolin-2 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:500-1:2000 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18672902
|
Novus Biologicals
NBP2-98731-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18609921
|
Novus Biologicals
NBP2-98731-50ul |
50 μg |
€ 224.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Caveolin-2 Polyclonal antibody specifically detects Caveolin-2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Caveolin-2 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Caveolae Markers, Membrane Trafficking and Chaperones, Neuroscience | |
| 0.2 um filtered solution in PBS | |
| 858 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry-Paraffin 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| CAV, caveolae protein, 20-kD, caveolin 2, caveolin 2 isoform a and b, caveolin-2, MGC12294 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human Caveolin-2. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title