missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caveolin-3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 326.00
Specifications
| Antigen | Caveolin-3 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:1000-1:5000 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18687882
|
Novus Biologicals
NBP2-98712-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18639811
|
Novus Biologicals
NBP2-98712-50ul |
50 μg |
N/A
|
N/A | |||||
Description
Caveolin-3 Polyclonal antibody specifically detects Caveolin-3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Caveolin-3 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Endocrinology, Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 859 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry-Paraffin 1:1000-1:5000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| caveolin 3, caveolin-3, LGMD1CMGC126100, LQT9MGC126101, M-caveolin, MGC126129, VIP21, VIP-21 | |
| Produced in rabbits immunized with A synthetic peptide corresponding to the N-terminus of the Human Caveolin-3. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title