Learn More
Invitrogen™ Caveolin 3 Recombinant Rabbit Monoclonal Antibody (23GB3035), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA553329
 View more versions of this product
Product Code. 30088569
  • € 207.00 / 20µL
Description
Caveolin-3 (CAV3) acts as a scaffolding protein within the caveolar membrane. It interacts directly with G-protein alpha subunits and can functionally regulate their activity. Caveolin-3 may also regulate voltage-gated potassium channels. It also plays a role in the sarcolemma repair mechanism of both skeletal muscle and cardiomyocytes that permits rapid resealing of membranes disrupted by mechanical stress. Caveolin-3 also mediates the recruitment of CAVIN2 and CAVIN3 proteins to the caveolae. Mutations in the gene can result in HyperCKmia, rippling muscle disease 2, cardiomyopathy, long QT syndrome 9, sudden infant death syndrome, and myopathy.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Caveolin 3
Recombinant Monoclonal
Unconjugated
Cav3
AI385751; CAV; Cav3; Cav-3; caveolin 3; caveolin-3; LGMD1C; LQT9; M-cav; M-caveolin; MGC126100; MGC126101; MGC126129; structural protein; VIP21; VIP-21
Rabbit
Affinity chromatography
RUO
29161, 859
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
23GB3035
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P51638, P56539
Cav3
A synthesized peptide derived from Caveolin-3 (1-60AA).
20 μL
Primary
Human, Rat