missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBARA1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-37983-20ul
This item is not returnable.
View return policy
Description
CBARA1 Polyclonal antibody specifically detects CBARA1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| CBARA1 | |
| Polyclonal | |
| Western Blot 1:100 - 1:500, ELISA | |
| Allergen Hom s 4, ara CALC, Atopy-related autoantigen CALC, CALCDKFZp564C246, calcium binding atopy-related autoantigen 1, Calcium-binding atopy-related autoantigen 1, MICU1mitochondrial, mitochondrial calcium uptake 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 105-255 of human CBARA1 (NP_001182447.1).,, Sequence:, GFRDRKVMEYENRIRAYSTPDKIFRYFATLKVISEPGEAEVFMTPEDFVRSITPNEKQPEHLGLDQYIIKRFDGKKISQEREKFADEGSIFYTLGECGLISFSDYIFLTTVLSTPQRNFEIAFKMFDLNGDGEVDMEEFEQVQSIIRSQTS | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10367 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu