missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBARA1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-37983-100ul
This item is not returnable.
View return policy
Description
CBARA1 Polyclonal antibody specifically detects CBARA1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| CBARA1 | |
| Polyclonal | |
| Western Blot 1:100 - 1:500, ELISA | |
| Allergen Hom s 4, ara CALC, Atopy-related autoantigen CALC, CALCDKFZp564C246, calcium binding atopy-related autoantigen 1, Calcium-binding atopy-related autoantigen 1, MICU1mitochondrial, mitochondrial calcium uptake 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 105-255 of human CBARA1 (NP_001182447.1).,, Sequence:, GFRDRKVMEYENRIRAYSTPDKIFRYFATLKVISEPGEAEVFMTPEDFVRSITPNEKQPEHLGLDQYIIKRFDGKKISQEREKFADEGSIFYTLGECGLISFSDYIFLTTVLSTPQRNFEIAFKMFDLNGDGEVDMEEFEQVQSIIRSQTS | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10367 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction