missing translation for 'onlineSavingsMsg'
Learn More

CBCP3 Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-98584R

Product Code. 18552783

  • € 415.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CBCP3 Polyclonal antibody specifically detects CBCP3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CBCP3
Polyclonal
50 mM Sodium Borate
Rabbit
Antigen and protein A Affinity-purified
RUO
340351
Store at 4°C in the dark.
IgG
Immunohistochemistry (Paraffin)
DyLight 550
ATP/GTP binding protein-like 3, ATP/GTP-binding protein-like 3, CCP3, cytosolic carboxypeptidase 3, EC 3.4.17, EC 3.4.17.-, FLJ12983, MGC32955
Produced in rabbits immunized with E. coli-derived Human CBCP3 fragment.
0.1 mL
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.