missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBFB Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-00328CL1
This item is not returnable.
View return policy
Description
CBFB Polyclonal antibody specifically detects CBFB in Human samples. It is validated for ELISA,Immunoprecipitation,Western Blot
Specifications
| CBFB | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunoprecipitation, Western Blot | |
| CoraFluor 1 | |
| CBF-beta, core-binding factor subunit beta, core-binding factor, beta subunit, PEA2-beta, PEBP2B, PEBP2-beta, polyomavirus enhancer binding protein 2, beta subunit, Polyomavirus enhancer-binding protein 2 beta subunit, SL3/AKV core-binding factor beta subunit, SL3-3 enhancer factor 1 beta subunit, SL3-3 enhancer factor 1 subunit beta | |
| Produced in rabbits immunized with purified, recombinant Human CBFB (Uniprot#: Q13951-1; Met1-Pro182) | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 865 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction