missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBFB Antibody (PCRP-CBFB-1F6) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14058JF525
This item is not returnable.
View return policy
Description
CBFB Monoclonal antibody specifically detects CBFB in Human samples. It is validated for Flow Cytometry,ELISA,Immunoprecipitation,Western Blot,Protein Array,Immunocytochemistry/ Immunofluorescence
Specifications
| CBFB | |
| Monoclonal | |
| Janelia Fluor 525 | |
| CBF-beta, core-binding factor subunit beta, core-binding factor, beta subunit, PEA2-beta, PEBP2B, PEBP2-beta, polyomavirus enhancer binding protein 2, beta subunit, Polyomavirus enhancer-binding protein 2 beta subunit, SL3/AKV core-binding factor beta subunit, SL3-3 enhancer factor 1 beta subunit, SL3-3 enhancer factor 1 subunit beta | |
| Recombinant full-length human CBFB protein (Uniprot: Q13951) | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 865 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, ELISA, Immunoprecipitation, Western Blot, Protein Array, Immunocytochemistry/Immunofluorescence | |
| PCRP-CBFB-1F6 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction