missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cbl-c Antibody (OTI3B4), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71755CL1
This item is not returnable.
View return policy
Description
Cbl-c Monoclonal antibody specifically detects Cbl-c in Human samples. It is validated for Flow Cytometry,Western Blot,CyTOF
Specifications
| Cbl-c | |
| Monoclonal | |
| CoraFluor 1 | |
| Cas-Br-M (murine) ecotropic retroviral transforming sequence c, Cas-Br-M (murine) ectropic retroviral transforming sequence c, CBL3, CBL-3, CBL-SL, RING finger protein 57, RNF57SH3-binding protein CBL-3, SH3-binding protein CBL-C, signal transduction protein CBL-C | |
| Human recombinant protein fragment corresponding to amino acids 244-474 of human CBLC(NP_036248) produced in E.coli. | |
| 0.1 mL | |
| Stem Cell Markers | |
| 23624 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| Flow Cytometry, Western Blot, CyTOF | |
| OTI3B4 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction