missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBL, p Tyr774 Antibody (SR1098), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | CBL |
|---|---|
| Clone | SR1098 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18694245
|
Novus Biologicals
NBP3-21601-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18620774
|
Novus Biologicals
NBP3-21601-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CBL Monoclonal antibody specifically detects CBL in Human samples. It is validated for Western Blot, Flow Cytometry, ImmunohistochemistrySpecifications
| CBL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Cas-Br-M (murine) ecotropic retroviral transforming sequence, Casitas B-lineage lymphoma proto-oncogene, CBL2E3 ubiquitin-protein ligase CBL, EC 6.3.2, EC 6.3.2.-, oncogene CBL2, Proto-oncogene c-Cbl, RING finger protein 55, RNF55C-CBL, Signal transduction protein CBL | |
| A synthesized peptide derived from human phospho-CBL [Tyr774] (Uniprot #: P22681) | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| SR1098 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Cancer, Phospho Specific, Signal Transduction, Zinc Finger | |
| PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
| 867 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title