missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CBMR 0083
GREENER_CHOICE

Product Code. 18774434
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18774434

Brand: Tocris Bioscience™ 7890/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent allosteric FFA1 (GPR40) and 4 (GPR120) agonist

CBMR 0083 is a potent allosteric free fatty acid receptor 1 (FFA1) and 4 (FFA4) agonist (EC50 values are 1 and 2 nM for FFA1 and FFA4, respectively).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-[3-[[4-Amino-6-[[2-(2,5-dichlorophenyl)ethyl]amino]-1,3,5-triazin-2-yl]methoxy]-5-methylphenyl]-2-pyrrolidinone
CAS 2839486-16-5
Quantity 10 mg
Target Free Fatty Acid Receptor Agonists
Molecular Formula C23H24Cl2N6O2
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.